Merck
An evolutionarily conserved pathway controls proteasome homeostasis.
Nature (2016-07-28)
Adrien Rousseau, Anne Bertolotti
ABSTRACT
The proteasome is essential for the selective degradation of most cellular proteins, but how cells maintain adequate amounts of proteasome is unclear. Here we show that there is an evolutionarily conserved signalling pathway controlling proteasome homeostasis. Central to this pathway is TORC1, the inhibition of which induced all known yeast 19S regulatory particle assembly-chaperones (RACs), as well as proteasome subunits. Downstream of TORC1 inhibition, the yeast mitogen-activated protein kinase, Mpk1, acts to increase the supply of RACs and proteasome subunits under challenging conditions in order to maintain proteasomal degradation and cell viability. This adaptive pathway was evolutionarily conserved, with mTOR and ERK5 controlling the levels of the four mammalian RACs and proteasome abundance. Thus, the central growth and stress controllers, TORC1 and Mpk1/ERK5, endow cells with a rapid and vital adaptive response to adjust proteasome abundance in response to the rising needs of cells. Enhancing this pathway may be a useful therapeutic approach for diseases resulting from impaired proteasomal degradation.
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-PSMD9 antibody produced in mouse, clone 3A4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ERK5 (big-MAPK, BMK1) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.