Merck
  • Copper-catalyzed enantioselective conjugate addition of dialkylzinc reagents to (2-pyridyl)sulfonyl imines of chalcones.
Copper-catalyzed enantioselective conjugate addition of dialkylzinc reagents to (2-pyridyl)sulfonyl imines of chalcones.
The Journal of organic chemistry (2005-08-27)
Jorge Esquivias, Ramon Gómez Arrayás, Juan C Carretero
ABSTRACT
[reaction: see text] The enantioselective catalytic 1,4-addition to alpha,beta-unsaturated ketimines is an unprecedented process. Herein, we document the copper-catalyzed addition of dialkylzinc reagents to (2-pyridylsulfonyl)imines of chalcones. This process occurs rapidly in the presence of a chiral phosphoramidite ligand to afford exclusively the 1,4-addition product. In the case of addition of dimethylzinc, enantioselectivities in the range 70-80% ee are obtained. The presence of the metal-coordinating 2-pyridylsulfonyl group proved to be essential for this reaction to proceed.
Sigma-Aldrich
(S,R,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine, 95% (HPLC)
Sigma-Aldrich
(S,S,S)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine, 97%