- miR-4299 mediates the invasive properties and tumorigenicity of human follicular thyroid carcinoma by targeting ST6GALNAC4.
miR-4299 mediates the invasive properties and tumorigenicity of human follicular thyroid carcinoma by targeting ST6GALNAC4.
Altered sialylation is closely associated with tumor progression and invasiveness. Micro-RNAs endogenous regulators of gene expression have been implicated in human thyroid carcinoma invasiveness. The objective of this study is to examine the alterations of miR-4299 and ST6GALNAC family in human follicular thyroid carcinoma during metastatic process. qRT-PCR showed the differential expressional profiles of miR-4299 and ST6GALNAC family in three kinds of thyroid cell lines (FTC-133,FTC-238, Nthy-ori 3-1) and clinical tissue specimens(malignant and borderline). The altered expression levels of ST6GALNAC4 were corresponding to invasive phenotypes of FTC-133 and FTC-238 cells both in vitro and in vivo. Further date indicated that miR-4299 regulated tumor progression and invasiveness by directly targeting ST6GALNAC4. This study implies the potential therapeutic application of miR-4299 and ST6GALNAC4 in modulating the invasion and tumorigenicity of follicular thyroid carcinoma cell.