Merck
  • Identification of FOXM1-induced epigenetic markers for head and neck squamous cell carcinomas.
Identification of FOXM1-induced epigenetic markers for head and neck squamous cell carcinomas.
Cancer (2013-10-12)
Sungjae Hwang, Swarna Mahadevan, Fatima Qadir, Iain L Hutchison, Daniela Elena Costea, Evelyn Neppelberg, Per Gunnar Liavaag, Ahmad Waseem, Muy-Teck Teh
ABSTRACT
Epigenetic reprogramming of the methylome has been implicated in all stages of cancer evolution. It is now well accepted that cancer cells exploit epigenetic reprogramming, a mechanism that regulates stem/progenitor cell renewal and differentiation, to promote cancer initiation and progression. The oncogene FOXM1 has been implicated in all major forms of human cancer. We have recently shown that aberrant upregulation of FOXM1 orchestrated a DNA methylation signature that mimics the cancer methylome landscape, from which we have identified a number of FOXM1-induced epigenetic markers. Differential promoter methylation and gene expression in clinical specimens were measured using commercially available bisulfite conversion kits and absolute quantitative PCR, respectively. Here, we investigated 8 FOXM1-induced differentially methylated genes, SPCS1, FLNA, CHPF, GLT8D1, C6orf136, MGAT1, NDUFA10, and PAFAH1B3, using human head and neck tissue specimens donated by 2 geographically independent patient cohorts from Norway and the United Kingdom. Two genes (GLT8D1 and C6orf136) were found to be differentially expressed in head and neck squamous cell carcinomas (HNSCCs). Using methylation-specific quantitative PCR, we confirmed that the promoters of GLT8D1 and C6orf136 were hypo- and hypermethylated, respectively, in HNSCC tissues. Given that epigenetic change precedes gene expression, methylation status of candidate genes identified from this study may represent a signature of premalignancy, rendering them potentially useful predictive biomarkers for precancer screening and/or therapeutic targets for cancer prevention.
Product Description
Roche
Proteinase K, recombinant, PCR Grade, Solution from Pichia pastoris