Merck
  • Sensing of 'danger signals' and pathogen-associated molecular patterns defines binary signaling pathways 'upstream' of Toll.
Sensing of 'danger signals' and pathogen-associated molecular patterns defines binary signaling pathways 'upstream' of Toll.
Nature immunology (2008-08-30)
Laure El Chamy, Vincent Leclerc, Isabelle Caldelari, Jean-Marc Reichhart
ABSTRACT
In drosophila, molecular determinants from fungi and Gram-positive bacteria are detected by circulating pattern-recognition receptors. Published findings suggest that such pattern-recognition receptors activate as-yet-unidentified serine-protease cascades that culminate in the cleavage of Spätzle, the endogenous Toll receptor ligand, and trigger the immune response. We demonstrate here that the protease Grass defines a common activation cascade for the detection of fungi and Gram-positive bacteria mediated by pattern-recognition receptors. The serine protease Persephone, shown before to be specific for fungal detection in a cascade activated by secreted fungal proteases, was also required for the sensing of proteases elicited by bacteria in the hemolymph. Hence, Persephone defines a parallel proteolytic cascade activated by 'danger signals' such as abnormal proteolytic activities.
Sigma-Aldrich
Protease from Bacillus sp., liquid, ≥16 U/g
Pricing and availability is not currently available.
Sigma-Aldrich
Protease from Aspergillus oryzae, ≥500 U/g
Pricing and availability is not currently available.