Merck
  • Differential Changes in Postsynaptic Density Proteins in Postmortem Huntington's Disease and Parkinson's Disease Human Brains.
Differential Changes in Postsynaptic Density Proteins in Postmortem Huntington's Disease and Parkinson's Disease Human Brains.
Journal of neurodegenerative diseases (2014-01-01)
C Fourie, E Kim, H Waldvogel, J M Wong, A McGregor, R L M Faull, J M Montgomery
ABSTRACT
NMDA and AMPA-type glutamate receptors and their bound membrane-associated guanylate kinases (MAGUKs) are critical for synapse development and plasticity. We hypothesised that these proteins may play a role in the changes in synapse function that occur in Huntington's disease (HD) and Parkinson's disease (PD). We performed immunohistochemical analysis of human postmortem brain tissue to examine changes in the expression of SAP97, PSD-95, GluA2 and GluN1 in human control, and HD- and PD-affected hippocampus and striatum. Significant increases in SAP97 and PSD-95 were observed in the HD and PD hippocampus, and PSD95 was downregulated in HD striatum. We observed a significant increase in GluN1 in the HD hippocampus and a decrease in GluA2 in HD and PD striatum. Parallel immunohistochemistry experiments in the YAC128 mouse model of HD showed no change in the expression levels of these synaptic proteins. Our human data show that major but different changes occur in glutamatergic proteins in HD versus PD human brains. Moreover, the changes in human HD brains differ from those occurring in the YAC128 HD mouse model, suggesting that unique changes occur at a subcellular level in the HD human hippocampus.
Sigma-Aldrich
Anti-DLG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse