Merck
  • Distribution and significance of interstitial fibrosis and stroma-infiltrating B cells in tongue squamous cell carcinoma.
Distribution and significance of interstitial fibrosis and stroma-infiltrating B cells in tongue squamous cell carcinoma.
Oncology letters (2016-03-22)
Xiao-Mei Lao, Yu-Jie Liang, Yu-Xiong Su, Si-En Zhang, X I Zhou, Gui-Qing Liao
ABSTRACT
Inflammation and desmoplasia are frequently identified in the tumor microenvironment, and have been demonstrated to be effective modulators of malignant biological events. However, the mechanisms by which the inflammatory microenvironment and interstitial fibrosis interact with one another remain to be elucidated. The present study aimed to investigate the degree of inflammation and interstitial fibrosis in tongue squamous cell carcinoma (TSCC), and how this acts to affect the outcome of TSCC. Tissue samples from 93 cases of TSCC and paired tumor-adjacent non-neoplastic tongue epithelium, as well as 14 cases of epithelial dysplasia, were used. Interstitial collagen fibers were assessed using Masson's trichrome stain. Immunohistochemical identification of cancer-associated fibroblasts (CAFs) and stroma-infiltrating B cells was performed via detection of α-smooth muscle actin (SMA), vimentin, desmin and cluster of differentiation 19 (CD19). The clinicopathological significance and overall survival of the TSCC patients were statistically analyzed. Regularly distributed CAFs and CD19
Sigma-Aldrich
MISSION® esiRNA, targeting human DES
Pricing and availability is not currently available.