Merck
  • Synthesis and Structure of NaZnSiO(3)OH, a New Chiral Zincosilicate Framework Material.
Synthesis and Structure of NaZnSiO(3)OH, a New Chiral Zincosilicate Framework Material.
Inorganic chemistry (2001-10-25)
A. M. Healey, M. T. Weller, A. R. Genge
ABSTRACT
The structure of NaZnSiO(3)OH, synthesized hydrothermally by reaction of Na(2)ZnSiO(4) and NaOH, has been determined from single-crystal X-ray and powder neutron diffraction data (orthorhombic, space group P2(1)2(1)2(1,) a = 7.6872(2) Å, b = 9.3899(2) Å, c = 5.155(1) Å, Z = 4). The structure has a framework constructed from ZnO(4) and SiO(3)OH tetrahedra with bridging and trigonal oxygen atoms. Channels formed from eight-membered rings of tetrahedra house the sodium ions and are also partially blocked by moderately strongly hydrogen bonded OH groups.
Sigma-Aldrich
Zinc oxide, 99.999% trace metals basis
Sigma-Aldrich
Zinc oxide, 99.99% trace metals basis
Sigma-Aldrich
Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Sodium metasilicate
Sigma-Aldrich
Sodium silicate solution, reagent grade