Merck
  • Rapid preparation of high-purity nuclear proteins from a small number of cultured cells for use in electrophoretic mobility shift assays.
Rapid preparation of high-purity nuclear proteins from a small number of cultured cells for use in electrophoretic mobility shift assays.
BMC immunology (2014-12-21)
Yuqian Luo, Takeshi Hara, Yuko Ishido, Aya Yoshihara, Kenzaburo Oda, Masahiko Makino, Norihisa Ishii, Naoki Hiroi, Koichi Suzuki
ABSTRACT
Highly purified nuclear protein is required when using an electrophoretic mobility shift assay (EMSA) to study transcription factors, e.g. nuclear factor-κB (NF-κB), a major transcription factor that regulates both innate and adaptive immune responses following infection. Although many protocols have been developed for nuclear protein extraction, they are not necessarily optimized for use in EMSA, often require a large number of cells and long processing times, and do not always result in complete separation of the nuclear and cytoplasmic fractions. We have developed a simple, rapid and cost-effective method to prepare highly purified nuclear proteins from a small number of both suspended and adherent cultured cells that yields nuclear proteins comparable to those prepared by a standard large-scale method. The efficiency of the method was demonstrated by using EMSA to show the successful detection, in multilple concurrent samples, of NF-κB activation upon tetradecanoyl phorbol acetate (TPA) stimulation. This method requires only a small number of cells and no specialized equipment. The steps have been simplified, resulting in a short processing time, which allows researchers to process multiple samples simultaneously and quickly. This method is especially optimized for use in EMSA, and may be useful for other applications that include proteomic analysis.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Pricing and availability is not currently available.