Merck
  • FGF17, a gene involved in cerebellar development, is downregulated in a patient with Dandy-Walker malformation carrying a de novo 8p deletion.
FGF17, a gene involved in cerebellar development, is downregulated in a patient with Dandy-Walker malformation carrying a de novo 8p deletion.
Neurogenetics (2011-04-13)
Ginevra Zanni, Sabina Barresi, Lorena Travaglini, Laura Bernardini, Teresa Rizza, Maria Cristina Digilio, Eugenio Mercuri, Stefano Cianfarani, Massimiliano Valeriani, Alessandro Ferraris, Letizia Da Sacco, Antonio Novelli, Enza Maria Valente, Bruno Dallapiccola, Enrico Silvio Bertini
ABSTRACT
Fibroblast growth factors (FGFs) are important signaling molecules which act during early vertebrate central nervous system development. FGF17, together with FGF8, is a key factor in the patterning of the mid-hindbrain region with a complex picture of spatiotemporal gene expression during the various stages of cerebellar development. Disruption or reduced expression of fgf17 in mice has been associated with cerebellar vermis abnormalities. We have identified a de novo 2.3-Mb deletion of chromosome 8p21.2-p21.3 in a girl with severe growth retardation, seizures, and classical Dandy-Walker malformation. Analysis of gene expression in blood lymphocytes and skin fibroblasts revealed markedly reduced levels of FGF17, which is located 1 Mb from the proximal deletion breakpoint. This is the first report of a human cerebellar malformation associated with transcriptional downregulation of the FGF17 gene.