Merck
  • Development of anti-HER2 fragment antibody conjugated to iron oxide nanoparticles for in vivo HER2-targeted photoacoustic tumor imaging.
Development of anti-HER2 fragment antibody conjugated to iron oxide nanoparticles for in vivo HER2-targeted photoacoustic tumor imaging.
Nanomedicine : nanotechnology, biology, and medicine (2015-08-05)
Kengo Kanazaki, Kohei Sano, Akira Makino, Yoichi Shimizu, Fumio Yamauchi, Satoshi Ogawa, Ning Ding, Tetsuya Yano, Takashi Temma, Masahiro Ono, Hideo Saji
ABSTRACT
Photoacoustic (PA) imaging is a promising imaging modality that provides biomedical information with high sensitivity and resolution. Iron oxide nanoparticles (IONPs) have been regarded as remarkable PA contrast agents because of their low toxicity and biodegradable properties. However, IONP delivery is restricted by its modest leakage and retention in tumors. In this study, we designed IONPs (20nm, 50nm, and 100nm) conjugated with anti-HER2 moieties [whole IgG, single-chain fragment variable (scFv), and peptide] for HER2-targeted PA tumor imaging. The binding affinity, cellular uptake, and in vivo biodistribution were examined. We propose 20-nm anti-HER2 scFv-conjugated IONPs (SNP20) as a novel PA contrast agent. SNP20 demonstrated high affinity and specific binding to HER2-expressing cells; it selectively visualized HER2-positive tumors in PA imaging studies. These data indicate that SNP20 is a potential PA contrast agent for imaging of HER2-expressing tumors. Iron oxide nanoparticles have been demonstrated to be good contrast agents for tumor imaging. They may also be useful in photoacoustic (PA) imaging, which can provide high sensitivity data and image resolution. The authors here coupled iron oxide nanoparticles with anti-HER2 antibody fragment and showed significant retention of these nanoparticles in tumors. This combination may provide another option for enhanced imaging of tumors.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
TNP, ≥95% (HPLC)
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%