Merck
  • Determination of manganese- and manganese-containing fungicides with lucigenin-Tween-20-enhanced chemiluminescence detection.
Determination of manganese- and manganese-containing fungicides with lucigenin-Tween-20-enhanced chemiluminescence detection.
Luminescence : the journal of biological and chemical luminescence (2015-02-03)
Mohammad Yaqoob, Mohammad Asghar, Abdul Nabi
ABSTRACT
A flow-injection (FI) method is reported for the determination of Mn(II), maneb and mancozeb fungicides based on the catalytic effect of Mn(II) on the oxidation of lucigenin and dissolved oxygen in a basic solution. The Tween-20 surfactant has been reported for first time to enhance lucigenin chemiluminescence (CL) intensity in the presence of Mn(II) (53%) and maneb and mancozeb (89%). The calibration graphs were linear in the concentration range of 0.001-1.5 mg L(-1) (R(2) = 0.9982 (n = 11) with a limit of detection (S/N = 3) of 0.1 µg L(-1) for Mn(II) and 0.01-3.0 mg L(-1) [R(2) = 0.9989 and R(2) = 0.9992 (n = 6)] with a limit of detection (S/N =3) of 1.0 µg L(-1) for maneb and mancozeb respectively. Injection throughputs of 90 and 120 h(-1) for Mn(II) and maneb and mancozeb respectively, and relative standard deviations of 1.0-3.4% were obtained in the concentration range studied. The experimental variables, e.g., reagents concentrations, flow rates, sample volume, and photomultiplier tube voltage, were optimized and potential interferences were investigated. The analysis of Mn(II) in river water reference materials (SLRS-4 and SLRS-5) showed good agreement with the certified values incorporating an on-line 8-hydroxyquinoline chelating column in the manifold for removing interfering metal ions. Recoveries for maneb and mancozeb were in the range of 92 ± 5 to 104 ± 3% and 91 ± 2 to 100 ± 4% (n = 3) respectively. The effect of 30 other pesticides (fungicides, herbicides and insecticides) was also examined in the lucigenin-Tween-20 CL system.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, 95%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Carbofuran, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis