Merck
  • FLT3-ITD confers resistance to the PI3K/Akt pathway inhibitors by protecting the mTOR/4EBP1/Mcl-1 pathway through STAT5 activation in acute myeloid leukemia.
FLT3-ITD confers resistance to the PI3K/Akt pathway inhibitors by protecting the mTOR/4EBP1/Mcl-1 pathway through STAT5 activation in acute myeloid leukemia.
Oncotarget (2015-04-01)
Ayako Nogami, Gaku Oshikawa, Keigo Okada, Shusaku Fukutake, Yoshihiro Umezawa, Toshikage Nagao, Tetsuya Kurosu, Osamu Miura
ABSTRACT
FLT3-ITD and FLT3-TKD are the most frequent tyrosine kinase mutations in acute myeloid leukemia (AML), with the former associated with poor prognosis. Here, we show that the PI3K inhibitor GDC-0941 or the Akt inhibitor MK-2206 induced apoptosis through the mitochondria-mediated intrinsic pathway more efficiently in hematopoietic 32D cells driven by FLT3-TKD (32D/TKD) than FLT3-ITD (32D/ITD), which robustly activated STAT5. The resistance to GDC-0941 and MK-2206 was gained by expression of the constitutively activated STAT5 mutant STAT5A1*6 in 32D/TKD cells, while it was abrogated by the STAT5 inhibitor pimozide in 32D/ITD cells or FLT3-ITD-expressing human leukemic MV4-11 cells. GDC-0941 or MK-2206 induced dephosphorylation of 4EBP1 more conspicuously in 32D/TKD than in 32D/ITD, which was prevented or augmented by STAT5A1*6 or pimozide, respectively, and correlated with downregulation of the eIF4E/eIF4G complex formation and Mcl-1 expression. Furthermore, exogenous expression of Mcl-1 endowed resistance to GDC-0941 and MK-2206 on 32D/TKD cells. Finally, it was confirmed in primary AML cells with FLT3-ITD that pimozide enhanced 4EBP1 dephosphorylation and Mcl-1 downregulation to augment cytotoxicity of GDC-0941. These data suggest that the robust STAT5 activation by FLT3-ITD protects cells treated with the PI3K/Akt pathway inhibitors from apoptosis by maintaining Mcl-1 expression through the mTORC1/4EBP1/eIF4E pathway.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Pimozide
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Rhein
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse