Merck
  • Effect of biomimetic zinc-containing tricalcium phosphate (Zn-TCP) on the growth and osteogenic differentiation of mesenchymal stem cells.
Effect of biomimetic zinc-containing tricalcium phosphate (Zn-TCP) on the growth and osteogenic differentiation of mesenchymal stem cells.
Journal of tissue engineering and regenerative medicine (2014-04-17)
Joshua Chou, Jia Hao, Hirokazu Hatoyama, Besim Ben-Nissan, Bruce Milthorpe, Makoto Otsuka
ABSTRACT
Several studies have shown the effectiveness of zinc-tricalcium phosphate (Zn-TCP) for bone tissue engineering. In this study, marine calcareous foraminifera possessing uniform pore size distribution were hydrothermally converted to Zn-TCP. The ability of a scaffold to combine effectively with mesenchymal stem cells (MSCs) is a key tissue-engineering aim. In order to demonstrate the osteogenic ability of MSCs with Zn-TCP, the scaffolds were cultured in an osteogenic induction medium to elicit an osteoblastic response. The physicochemical properties of Zn-TCP were characterized by XRD, FT-IR and ICP-MS. MSCs were aspirated from rat femurs and cultured for 3 days before indirectly placing four samples into each respective well. After culture for 7, 10 and 14 days, osteoblastic differentiation was evaluated using alizarin red S stain, measurement of alkaline phosphatase (ALP) levels, cell numbers and cell viability. XRD and FT-IR patterns both showed the replacement of CO(3)(2-) with PO(4)(3-). Chemical analysis showed zinc incorporation of 5 mol%. Significant increases in cell numbers were observed at 10 and 14 days in the Zn-TCP group, while maintaining high levels of cell viability (> 90%). ALP activity in the Zn-TCP group was statistically higher at 10 days. Alizarin red S staining also showed significantly higher levels of calcium mineralization in Zn-TCP compared with the control groups. This study showed that MSCs in the presence of biomimetically derived Zn-TCP can accelerate their differentiation to osteoblasts and could potentially be useful as a scaffold for bone tissue engineering.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate, 96%