Merck
  • Low concentrations of Bisphenol A and para-Nonylphenol affect extravillous pathway of human trophoblast cells.
Low concentrations of Bisphenol A and para-Nonylphenol affect extravillous pathway of human trophoblast cells.
Molecular and cellular endocrinology (2015-06-02)
Antonella Spagnoletti, Luana Paulesu, Chiara Mannelli, Leonardo Ermini, Roberta Romagnoli, Marcella Cintorino, Francesca Ietta
ABSTRACT
Bisphenol A (BPA) and para-Nonylphenol (p-NP) are chemicals of industrial origin which may influence human reproductive health. The effects of these substances in the prenatal life is an important topic that is receiving greater attention in the developed countries. In this study, human trophoblast cells HTR-8/SVneo were exposed to BPA and p-NP (1 × 10(-15), 1 × 10(-13), 1 × 10(-11), 1 × 10(-9) and 1 × 10(-7) M) and incubated for 24, 48 and/or 72 h then, examined for the main physiological processes which characterize the extravillous trophoblast. Cell proliferation showed no changes while the processes of cell migration and invasion were both reduced by BPA and p-NP. For each chemical, the activity was higher at lower concentrations with a maximum activity between 1 × 10(-13) and 1 × 10(-11) M (p < 0.05 for 1 × 10(-9) and p < 0.001 for 1 × 10(-11) M). Co-culture studies with human umbilical cord endothelial cells (HUVEC) revealed that trophoblast/endothelial interaction was significantly reduced by p-NP at 1 × 10(-11) M. Moreover, both chemicals were inducing differentiation of HTR-8/SVneo toward polyploidy by the process of endoreduplication. The estrogen-receptor antagonist ICI significantly reduced p-NP action, while it had no effect on BPA treated cells. In conclusion, p-NP and BPA act on trophoblast cells altering key physiological processes in placenta development. The exact mechanism of action of the chemicals in human trophoblast still needs to be clarified.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Benzenesulfonyl fluoride, 99%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
E-64, protease inhibitor
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)