Merck
  • Chromatographic analysis of olopatadine in hydrophilic interaction liquid chromatography.
Chromatographic analysis of olopatadine in hydrophilic interaction liquid chromatography.
Journal of chromatographic science (2014-10-11)
Jelena Maksić, Marko Jovanović, Tijana Rakić, Igor Popović, Darko Ivanović, Biljana Jančić-Stojanović
ABSTRACT
In this paper, chromatographic analysis of active substance olopatadine hydrochloride, which is used in eye drops as antihistaminic agent, and its impurity E isomer by hydrophilic interaction liquid chromatography (HILIC) and application of design of experiments (DoE) methodology are presented. In addition, benzalkonium chloride is very often used as a preservative in eye drops. Therefore, the evaluation of its chromatographic behavior in HILIC was carried out as well. In order to estimate chromatographic behavior and set optimal chromatographic conditions, DoE methodology was applied. After the selection of important chromatographic factors, Box-Behnken design was utilized, and on the basis of the obtained models factor effects were examined. Then, multi-objective robust optimization is performed aiming to obtain chromatographic conditions that comply with several quality criteria simultaneously: adequate and robust separation of critical peak pair and maximum retention of the first eluting peak. The optimal conditions are identified by using grid point search methodology. The experimental verification confirmed the adequacy of the defined optimal conditions. Finally, under optimal chromatographic conditions, the method was validated and applicability of the proposed method was confirmed.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Olopatadine hydrochloride, ≥98% (HPLC)