Merck
  • The SWI/SNF ATPases Are Required for Triple Negative Breast Cancer Cell Proliferation.
The SWI/SNF ATPases Are Required for Triple Negative Breast Cancer Cell Proliferation.
Journal of cellular physiology (2015-03-27)
Qiong Wu, Pasil Madany, Jacqueline Akech, Jason R Dobson, Stephen Douthwright, Gillian Browne, Jennifer L Colby, Georg E Winter, James E Bradner, Jitesh Pratap, Greenfield Sluder, Rohit Bhargava, Simion I Chiosea, Andre J van Wijnen, Janet L Stein, Gary S Stein, Jane B Lian, Jeffrey A Nickerson, Anthony N Imbalzano
ABSTRACT
The Brahma (BRM) and Brahma-related Gene 1 (BRG1) ATPases are highly conserved homologs that catalyze the chromatin remodeling functions of the multi-subunit human SWI/SNF chromatin remodeling enzymes in a mutually exclusive manner. SWI/SNF enzyme subunits are mutated or missing in many cancer types, but are overexpressed without apparent mutation in other cancers. Here, we report that both BRG1 and BRM are overexpressed in most primary breast cancers independent of the tumor's receptor status. Knockdown of either ATPase in a triple negative breast cancer cell line reduced tumor formation in vivo and cell proliferation in vitro. Fewer cells in S phase and an extended cell cycle progression time were observed without any indication of apoptosis, senescence, or alterations in migration or attachment properties. Combined knockdown of BRM and BRG1 showed additive effects in the reduction of cell proliferation and time required for completion of cell cycle, suggesting that these enzymes promote cell cycle progression through independent mechanisms. Knockout of BRG1 or BRM using CRISPR/Cas9 technology resulted in the loss of viability, consistent with a requirement for both enzymes in triple negative breast cancer cells.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, BioUltra, ≥99%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human SMARCA2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Smarca2