Merck
Rac1/Pak1/p38/MMP-2 Axis Regulates Angiogenesis in Ovarian Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-01-18)
Vianey Gonzalez-Villasana, Enrique Fuentes-Mattei, Cristina Ivan, Heather J Dalton, Cristian Rodriguez-Aguayo, Ricardo J Fernandez-de Thomas, Burcu Aslan, Paloma Del C Monroig, Guermarie Velazquez-Torres, Rebecca A Previs, Sunila Pradeep, Nermin Kahraman, Huamin Wang, Pinar Kanlikilicer, Bulent Ozpolat, George Calin, Anil K Sood, Gabriel Lopez-Berestein
ABSTRACT
Zoledronic acid is being increasingly recognized for its antitumor properties, but the underlying functions are not well understood. In this study, we hypothesized that zoledronic acid inhibits ovarian cancer angiogenesis preventing Rac1 activation. The biologic effects of zoledronic acid were examined using a series of in vitro [cell invasion, cytokine production, Rac1 activation, reverse-phase protein array, and in vivo (orthotopic mouse models)] experiments. There was significant inhibition of ovarian cancer (HeyA8-MDR and OVCAR-5) cell invasion as well as reduced production of proangiogenic cytokines in response to zoledronic acid treatment. Furthermore, zoledronic acid inactivated Rac1 and decreased the levels of Pak1/p38/matrix metalloproteinase-2 in ovarian cancer cells. In vivo, zoledronic acid reduced tumor growth, angiogenesis, and cell proliferation and inactivated Rac1 in both HeyA8-MDR and OVCAR-5 models. These in vivo antitumor effects were enhanced in both models when zoledronic acid was combined with nab-paclitaxel. Zoledronic acid has robust antitumor and antiangiogenic activity and merits further clinical development as ovarian cancer treatment.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human RAC1
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rac1