Merck
  • Protective Effects of Lactobacillus plantarum CCFM8246 against Copper Toxicity in Mice.
Protective Effects of Lactobacillus plantarum CCFM8246 against Copper Toxicity in Mice.
PloS one (2015-11-26)
Fengwei Tian, Yue Xiao, Xiaoxiao Li, Qixiao Zhai, Gang Wang, Qiuxiang Zhang, Hao Zhang, Wei Chen
ABSTRACT
Lactobacillus plantarum CCFM8246, which has a relatively strong copper binding capacity and tolerance to copper ions, was obtained by screening from 16 lactic acid bacteria in vitro. The selected strain was then applied to a mouse model to evaluate its protective function against copper intoxication in vivo. The experimental mice were divided into an intervention group and a therapy group; mice in the intervention group received co-administration of CCFM8246 and a copper ion solution by gavage, while mice in the therapy group were treated with CCFM8246 after 4 weeks of copper exposure. In both two groups, mice treated with copper alone and that treated with neither CCFM8246 nor copper served as positive and negative controls, respectively. At the end of the experimental period, the copper content in feces and tissues, the activity of alanine aminotransferase (ALT) and aspartate aminotransferase (AST) in serum, and oxidation stress indices in liver and kidney tissue were determined. Learning and memory ability was evaluated by Morris water maze experiments. The results indicated that treatment with CCFM8246 significantly increased the copper content in feces to promote copper excretion, reduce the accumulation of copper in tissues, reverse oxidative stress induced by copper exposure, recover the ALT and AST in serum and improve the spatial memory of mice.
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)