Merck
  • Altered metabolism of LDL in the arterial wall precedes atherosclerosis regression.
Altered metabolism of LDL in the arterial wall precedes atherosclerosis regression.
Circulation research (2015-09-12)
Emil D Bartels, Christina Christoffersen, Marie W Lindholm, Lars B Nielsen
ABSTRACT
Plasma cholesterol lowering is beneficial in patients with atherosclerosis. However, it is unknown how it affects entry and degradation of low-density lipoprotein (LDL) particles in the lesioned arterial wall. We studied the effect of lipid-lowering therapy on LDL permeability and degradation of LDL particles in atherosclerotic aortas of mice by measuring the accumulation of iodinated LDL particles in the arterial wall. Cholesterol-fed, LDL receptor-deficient mice were treated with either an anti-Apob antisense oligonucleotide or a mismatch control antisense oligonucleotide once a week for 1 or 4 weeks before injection with preparations of iodinated LDL particles. The anti-Apob antisense oligonucleotide reduced plasma cholesterol by ≈90%. The aortic LDL permeability and degradation rates of newly entered LDL particles were reduced by ≈50% and ≈85% already after 1 week of treatment despite an unchanged pool size of aortic iodinated LDL particles. In contrast, the size, foam cell content, and aortic pool size of iodinated LDL particles of aortic atherosclerotic plaques were not reduced until after 4 weeks of treatment with the anti-Apob antisense oligonucleotide. Improved endothelial barrier function toward the entry of plasma LDL particles and diminished aortic degradation of the newly entered LDL particles precede plaque regression.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Tyramine, 98%, FG
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Tyramine, ≥98.0%
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, 99.999% trace metals basis