Merck
  • A set of powerful negative selection systems for unmodified Enterobacteriaceae.
A set of powerful negative selection systems for unmodified Enterobacteriaceae.
Nucleic acids research (2015-03-25)
Varnica Khetrapal, Kurosh Mehershahi, Shazmina Rafee, Siyi Chen, Chiew Ling Lim, Swaine L Chen
ABSTRACT
Creation of defined genetic mutations is a powerful method for dissecting mechanisms of bacterial disease; however, many genetic tools are only developed for laboratory strains. We have designed a modular and general negative selection strategy based on inducible toxins that provides high selection stringency in clinical Escherichia coli and Salmonella isolates. No strain- or species-specific optimization is needed, yet this system achieves better selection stringency than all previously reported negative selection systems usable in unmodified E. coli strains. The high stringency enables use of negative instead of positive selection in phage-mediated generalized transduction and also allows transfer of alleles between arbitrary strains of E. coli without requiring phage. The modular design should also allow further extension to other bacteria. This negative selection system thus overcomes disadvantages of existing systems, enabling definitive genetic experiments in both lab and clinical isolates of E. coli and other Enterobacteriaceae.
Sigma-Aldrich
Anti-GroEL antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood