Merck
  • β2-Adrenoreceptor-Mediated Proliferation Inhibition of Embryonic Pluripotent Stem Cells.
β2-Adrenoreceptor-Mediated Proliferation Inhibition of Embryonic Pluripotent Stem Cells.
Journal of cellular physiology (2015-02-03)
Fan Sun, Xin-Jie Yang, Hao-Yu Lv, Ya-Bin Tang, Shi-Min An, Xu-Ping Ding, Wen-Bin Li, Lin Teng, Ying Shen, Hong-Zhuan Chen, Liang Zhu
ABSTRACT
Adrenoreceptors (ARs) are widely expressed and play essential roles throughout the body. Different subtype adrenoceptors elicit distinct effects on cell proliferation, but knowledge remains scarce about the subtype-specific effects of β2-ARs on the proliferation of embryonic pluripotent stem (PS) cells that represent different characteristics of proliferation and cell cycle regulation with the somatic cells. Herein, we identified a β2-AR/AC/cAMP/PKA signaling pathway in embryonic PS cells and found that the pathway stimulation inhibited proliferation and cell cycle progression involving modulating the stem cell growth and cycle regulatory machinery. Embryonic stem (ES) cells and embryonal carcinoma stem (ECS) cells expressed functional β-ARs coupled to AC/cAMP/PKA signaling. Agonistic activation of β-ARs led to embryonic PS cell cycle arrest and proliferation inhibition. Pharmacological and genetic analyzes using receptor subtype blocking and RNA interference approaches revealed that this effect selectively depended on β2-AR signaling involving the regulation of AKT, ERK, Rb, and Cyclin E molecules. Better understanding of the effects of β2-ARs on embryonic PS cell proliferation and cycle progression may provide new insights into stem cell biology and afford the opportunity for exploiting more selective ligands targeting the receptor subtype for the modulation of stem cells.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b7
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b8
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akr1b3
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting human AREG
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fdxr
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Areg
Sigma-Aldrich
MISSION® esiRNA, targeting human AR
Sigma-Aldrich
MISSION® esiRNA, targeting human FDXR
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
SAFC
L-Glutamine
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
MISSION® esiRNA, targeting human AKR1B1
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture