Merck
  • Haemocompatibility and cytotoxic studies of non-metallic composite materials modified with magnetic nano and microparticles.
Haemocompatibility and cytotoxic studies of non-metallic composite materials modified with magnetic nano and microparticles.
Acta of bioengineering and biomechanics (2015-12-22)
Maria Szymonowicz, Zbigniew Rybak, Aneta Fraczek-Szczypta, Danuta Paluch, Agnieszka Rusak, Katarzyna Nowicka, Marta Blazewicz
ABSTRACT
Preventing the formation of blood clots on the surface of biomaterials and investigation of the reasons of their formation are the leading topics of the research and development of biomaterials for implants placed into the bloodstream. Biocompatibility and stability of a material in body fluids and direct effect on blood cell counts components are related both to the structure and physico-chemical state of an implant surface. The aim of this study was to determine haemocompatibility and cytotoxicity of polysulfone-based samples containing nano and micro particles of magnetite (Fe3O4). The polysulfone-based samples modified with nanometric and micrometric magnetite particles were examined. Physicochemical properties of the composites were determined by testing their wettability and surface roughness. The action of haemolytic, activation of coagulation system and cytotoxicity of composites was evaluated. Wettability and roughness of materials were correlated with nanoparticles and microparticles content. In the tests of plasma coagulation system shortening of activated partial thromboplastin time for polysulfone with nano magnetite and with micro magnetite particles was observed in comparison with pure polysulfone. Prothrombine time and thrombine time values as well as fibrinogen concentration were unchanged. Haemolysis values were normal. Morphology and viability of cells were normal. Composites made from polysulfone modified with nanoparticles and microparticles of magnetite cause neither haemolytic nor cytotoxic reaction. These composites evoke plasma endogenous system activation.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
SAFC
L-Glutamine
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture