Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.0125mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Manganese, rod, 50mm, diameter 4.0mm, cast, 99.5%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
SKU
Pack Size
Availability
Price
Quantity
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%