Merck
CD244 is expressed on dendritic cells and regulates their functions.
Immunology and cell biology (2015-02-04)
Anna-Maria Georgoudaki, Sorosh Khodabandeh, Speranta Puiac, Catrine M Persson, Maria K Larsson, Max Lind, Oscar Hammarfjord, Tara H Nabatti, Robert P A Wallin, Ulf Yrlid, Mikael Rhen, Vinay Kumar, Benedict J Chambers
ABSTRACT
Signaling lymphocytic activation molecule (SLAM) receptors have an important role in the development of immune responses because of their roles, for exampe, in NK cell cytotoxicity and cytokine production by NK, T cells and myeloid cells. The SLAM receptor CD244 (2B4, SLAMf4) is expressed on a variety of immune cell types but most of its functions have been examined on NK and T cells. In the present study, we investigated expression and function of CD244 in murine subsets of dendritic cells (DCs). We report that all subsets of murine DCs examined expressed CD244, although the expression levels of CD244 varied between subsets. Splenic and resident mesenteric lymph node (MLN) DCs from CD244(-/-) mice expressed lower levels of CD86 and MHC class II compared with wild-type mice. Upon Toll-like receptor (TLR) stimulation, no differences in surface expression of these molecules were observed between DCs from CD244(-/-) and wild-type mice. However, splenic DCs from CD244(-/-) mice upon stimulation with TLR binding ligands lipopolysaccharide (LPS) and CpG produced significantly higher levels of pro-inflammatory cytokines. In addition, DCs from CD244(-/-) mice elicited increased NK cell activation in vitro. These data add CD244 to a growing list of immuno-modulatory receptors found on DCs.
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)