Merck
  • Structure design and manufacturing of layered bioceramic scaffolds for load-bearing bone reconstruction.
Structure design and manufacturing of layered bioceramic scaffolds for load-bearing bone reconstruction.
Biomedical materials (Bristol, England) (2015-07-15)
Jing-Zhou Yang, Xiao-Zhi Hu, Rumana Sultana, Robert Edward Day, Paul Ichim
ABSTRACT
Bioceramic scaffolds with desired bone regeneration functions have the potential to become real alternatives to autologous bone grafts for reconstruction of load-bearing and critical-sized segmental bone defects. The aim of this paper was to develop a layered scaffold structure that has the biodegradable function of common monolithic scaffolds and adequate mechanical function for surgical fixing and after surgery support. The exemplary case of this study is assumed to be a large-segment tibia or femur bone repair. The layered scaffold structure consists of a macro porous hydroxyapatite-wollastonite layer and a strong dense zirconia matrix dense layer. The bio-functional scaffold layer with interconnected freeze-dried porous structures shows excellent apatite formation, cell attachment, and cell proliferation capabilities. The mechanical functional layer provides a bending strength matching that of the compact bone.
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O