Merck
The human decapping scavenger enzyme DcpS modulates microRNA turnover.
Scientific reports (2015-11-21)
Oussama Meziane, Sandra Piquet, Gabriel D Bossé, Dominic Gagné, Eric Paquet, Claude Robert, Michael A Tones, Martin J Simard
ABSTRACT
The decapping scavenger enzyme DcpS is known for its role in hydrolyzing the cap structure following mRNA degradation. Recently, we discovered a new function in miRNA degradation activation for the ortholog of DcpS in C. elegans. Here we show that human DcpS conserves its role in miRNA turnover. In human cells, DcpS is a nucleocytoplasmic shuttling protein that activates miRNA degradation independently of its scavenger decapping activity in the cytoplasmic compartment. We also demonstrate that this new function for DcpS requires the contribution of the 5'-3' exonuclease Xrn2. Our findings support a conserved role of DcpS as a modulator of miRNA turnover in animals.
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
MISSION® esiRNA, targeting human XRN1
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Xrn2
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Xrn1
SAFC
HEPES
Sigma-Aldrich
MISSION® esiRNA, targeting human XRN2
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%