Merck
  • MRP1 knockdown down-regulates the deposition of collagen and leads to a reduced hypertrophic scar fibrosis.
MRP1 knockdown down-regulates the deposition of collagen and leads to a reduced hypertrophic scar fibrosis.
Journal of molecular histology (2015-06-21)
Yan Li, Longlong Yang, Zhao Zheng, Jihong Shi, Xue Wu, Hao Guan, Yanhui Jia, Ke Tao, Hongtao Wang, Shichao Han, Jianxin Gao, Bin Zhao, Linlin Su, Dahai Hu
ABSTRACT
Multidrug resistance-associated protein 1 (MRP1) belongs to ATP-binding cassette transporters family. The overexpression of MRP1 is predominantly related with the failure of chemo-radiotherapy in various tumors. However, its possible role in hypertrophic scar (HS) is hardly investigated. Here we showed that the mRNA level and protein expression of MRP1 were higher in HS and HS derived fibroblasts (HSFs) than that in normal skin (NS) and NS derived fibroblasts (NSFs). Immunohistochemistry and immunofluorescence showed that the percentage of positive cells was higher in HS and HSFs. Meanwhile, the co-localization of MRP1 and α-SMA was stronger in HS. MRP1 knockdown in HSFs provoked a significant reduction in the protein expressions of collagen 3 and α-SMA in vitro. Moreover, MRP1 siRNA transfection could decrease the deposition of collagen in cultured tissues ex vivo and inhibit the scar formation in rabbit ear scar model in vivo. H&E staining and Masson trichrome staining revealed thinner and more orderly arranged collagen fiber in the MRP1 siRNA transfection group. The appearance of scar was improved as well. All these results indicate that MRP1 plays an important role in the formation of HS, MRP1 knockdown could be a potential method to reduce the accumulation of collagen and to improve the abnormal deposition of extracellular matrix in HS, which indicates that down-regulation of MRP1 has the potential therapeutic effect in the treatment and prophylaxis of HS.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Abcc1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mdm4
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting human MSH3
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Msh3
Sigma-Aldrich
MISSION® esiRNA, targeting human MDM4
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC1