Merck
  • Insulin B chain 9-23 gene transfer to hepatocytes protects from type 1 diabetes by inducing Ag-specific FoxP3+ Tregs.
Insulin B chain 9-23 gene transfer to hepatocytes protects from type 1 diabetes by inducing Ag-specific FoxP3+ Tregs.
Science translational medicine (2015-05-29)
Mahzad Akbarpour, Kevin S Goudy, Alessio Cantore, Fabio Russo, Francesca Sanvito, Luigi Naldini, Andrea Annoni, Maria Grazia Roncarolo
ABSTRACT
Antigen (Ag)-specific tolerance in type 1 diabetes (T1D) in human has not been achieved yet. Targeting lentiviral vector (LV)-mediated gene expression to hepatocytes induces active tolerance toward the encoded Ag. The insulin B chain 9-23 (InsB9-23) is an immunodominant T cell epitope in nonobese diabetic (NOD) mice. To determine whether auto-Ag gene transfer to hepatocytes induces tolerance and control of T1D, NOD mice were treated with integrase-competent LVs (ICLVs) that selectively target the expression of InsB9-23 to hepatocytes. ICLV treatment induced InsB9-23-specific effector T cells but also FoxP3(+) regulatory T cells (Tregs), which halted islet immune cell infiltration, and protected from T1D. Moreover, ICLV treatment combined with a single suboptimal dose of anti-CD3 monoclonal antibody (mAb) is effective in T1D reversal. Splenocytes from LV.InsB9-23-treated mice, but not from LV.OVA (ovalbumin)-treated control mice, stopped diabetes development, demonstrating that protection is Ag-specific. Depletion of CD4(+)CD25(+)FoxP3(+) T cells led to diabetes progression, indicating that Ag-specific FoxP3(+) Tregs mediate protection. Integrase-defective LVs (IDLVs).InsB9-23, which alleviate the concerns for insertional mutagenesis and support transient transgene expression in hepatocytes, were also efficient in protecting from T1D. These data demonstrate that hepatocyte-targeted auto-Ag gene expression prevents and resolves T1D and that stable integration of the transgene is not required for this protection. Gene transfer to hepatocytes can be used to induce Ag-specific tolerance in autoimmune diseases.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications