Merck
  • The DRBD13 RNA binding protein is involved in the insect-stage differentiation process of Trypanosoma brucei.
The DRBD13 RNA binding protein is involved in the insect-stage differentiation process of Trypanosoma brucei.
FEBS letters (2015-06-02)
Bhaskar Anand Jha, Vahid H Gazestani, Chun Wai Yip, Reza Salavati
ABSTRACT
DRBD13 RNA-binding protein (RBP) regulates the abundance of AU-rich element (ARE)-containing transcripts in trypanosomes. Here we show that DRBD13 regulates RBP6, the developmentally critical protein in trypanosomatids. We also show DRBD13-specific regulation of transcripts encoding cell surface coat proteins including GPEET2, variable surface glycoprotein (VSG) and invariant surface glycoprotein (ISG). Accordingly, alteration in DRBD13 levels leads to changes in the target mRNA abundance and parasite morphology. The high consistency of the observed phenotype with known cell membrane exchanges that occur during progression of T. brucei through the insect stage of its life cycle suggests that DRBD13 is an important regulator in this largely unknown developmental process.
Sigma-Aldrich
Anti-Chloramphenicol Acetyl Transferase (CAT) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution