Merck
  • The potential of DBP gels containing intervertebral disc cells for annulus fibrosus supplementation: in vivo.
The potential of DBP gels containing intervertebral disc cells for annulus fibrosus supplementation: in vivo.
Journal of tissue engineering and regenerative medicine (2013-03-06)
Jeong Eun Song, Eun Young Kim, Woo Young Ahn, Yu Jeong Lee, Dongwon Lee, Rui Reis, Gilson Khang
ABSTRACT
Demineralized bone particle (DBP), which is widely used as a biomaterial in the field of tissue engineering, contains various bioactive molecules, such as cytokines. For this reason, in this study we investigated the effects of injectable DBP gels on cell proliferation, inflammation and maintenance of the shape of DBP gels as a scaffold able to substitute for intervertebral discs (IVDs) in vivo. DBP gels were fabricated with different percentages (5% and 10%) of DBP powder and 3% acetic acid, including 0.02% pepsin. DBP gels with 1 × 10(6) annulus fibrosus (AF) cells were implanted into the dorsal subcutaneous region of BALB/C-nu mice for 1, 2 and 3 weeks. Cell proliferation was measured by MTT assay. The effect of DBP gels on the inflammatory response was analysed by measuring the amount of tumour necrosis factor-alpha (TNFα) released. Also, histological methods were carried out to analyse the response of DBP gels in vivo. This study demonstrated that injectable DBP gels are able to provide physical scaffolds for growing IVD cells in vivo.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%