Merck
  • Structure-Based Design of Macrocyclic Coagulation Factor VIIa Inhibitors.
Structure-Based Design of Macrocyclic Coagulation Factor VIIa Inhibitors.
Journal of medicinal chemistry (2015-07-08)
E Scott Priestley, Daniel L Cheney, Indawati DeLucca, Anzhi Wei, Joseph M Luettgen, Alan R Rendina, Pancras C Wong, Ruth R Wexler
ABSTRACT
On the basis of a crystal structure of a phenylpyrrolidine lead and subsequent molecular modeling results, we designed and synthesized a novel series of macrocyclic FVIIa inhibitors. The optimal 16-membered macrocycle was 60-fold more potent than an acyclic analog. Further potency optimization by incorporation of P1' alkyl sulfone and P2 methyl groups provided a macrocycle with TF/FVIIa Ki = 1.6 nM, excellent selectivity against a panel of seven serine proteases, and FVII-deficient prothrombin time EC2x = 1.2 μM. Discovery of this potent, selective macrocyclic scaffold opens new possibilities for the development of orally bioavailable FVIIa inhibitors.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O
Sigma-Aldrich
Hydroxylamine solution, 50 wt. % in H2O, 99.999%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications