Merck
  • Transcription factors E2F1 and E2F3 are expressed in placenta but do not regulate MMP14.
Transcription factors E2F1 and E2F3 are expressed in placenta but do not regulate MMP14.
Placenta (2015-07-07)
T J Kaitu'u-Lino, R Hastie, P Cannon, H Nguyen, S Lee, N J Hannan, S Tong
ABSTRACT
Preeclampsia is a serious complication of pregnancy for which there are no efficacious medical treatments. Soluble endoglin is as an anti-angiogenic factor that contributes to the pathogenesis of the disease, however little is known about its molecular regulation in placenta. Recent data has demonstrated E2F transcription factors directly regulate MMPs in metastatic disease. Of particular interest was the capacity of E2F1 and E2F3 to up-regulate MMP14, a protease that cleaves and releases soluble endoglin from placenta. The aim of this study was to characterize E2F1 and E2F3 in preeclamptic placenta and assess whether silencing affects soluble endoglin release. E2F1 and E2F3 mRNA, protein expression and localization were assessed in severe early onset preeclamptic and preterm control placentas (delivered <34 weeks gestation). E2F siRNA was administered to primary trophoblast and primary endothelial cells and effect on MMP14 mRNA expression and soluble endoglin secretion assessed. E2F1 and E2F3 were localized to the syncytiotrophoblast. E2F1 was significantly down regulated in severe preeclamptic placentas, while E2F3 was unchanged. Silencing E2F1 did not decrease MMP14 expression in primary trophoblast or endothelial cells. However, E2F1 silencing resulted in a significant increase in soluble endoglin secretion from both cell types, and silencing of E2F3 also significantly increased soluble endoglin release from primary trophoblast. This study demonstrates that E2F1 and E2F3 are present within the syncytiotrophoblast of placenta and that E2F1 is reduced in preeclampsia. Although silencing of either E2F1 or E2F3 does not alter MMP14 expression, both appear to regulate soluble endoglin release.
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
MISSION® esiRNA, targeting human E2F1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse E2f1