Merck
  • The role of TXNDC5 in castration-resistant prostate cancer-involvement of androgen receptor signaling pathway.
The role of TXNDC5 in castration-resistant prostate cancer-involvement of androgen receptor signaling pathway.
Oncogene (2014-12-17)
L Wang, G Song, X Chang, W Tan, J Pan, X Zhu, Z Liu, M Qi, J Yu, B Han
ABSTRACT
Castration-resistant prostate cancer (CRPC) continues to be a major clinical problem and the mechanisms behind it remain unclear. Thioredoxin domain-containing protein 5 (TXNDC5) is involved in protein folding and chaperone activity, and its overexpression has been reported in multiple malignancies. In the current study, we demonstrated that TXNDC5 is up-regulated following long-term androgen-deprivation treatment (ADT) and is highly overexpressed in CRPC tumors compared with hormone-naive prostate cancer (PCa) cases. Functionally, in vitro and in vivo studies demonstrated that TXNDC5 overexpression promotes the growth of both androgen-dependent and castration-resistant PCa xenografts. Mechanistically, TXNDC5 directly interacts with the AR protein to increase its stability and thus enhances its transcriptional activity. TXDNC5-mediated CRPC growth can be fully abolished by AR inhibition, suggesting TXDNC5 up-regulation as an escape pathway for aberrant AR re-activation and CRPC growth in the milieu of low androgen. Indeed, we found that TXNDC5 is increased by ADT-induced hypoxia through HIF-1α in an miR-200b-dependent manner. Overall, we defined an important role of TXNDC5 in CRPC and further investigations are needed to screen TXNDC5 antagonists as a novel therapeutic approaches to treat PCa patients with CRPC.
Sigma-Aldrich
MISSION® esiRNA, targeting human BLOC1S5
Sigma-Aldrich
MISSION® esiRNA, targeting human TXNDC5, BLOC1S5-TXNDC5
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Txndc5