Merck
  • Development of highly stabilized curcumin nanoparticles by flash nanoprecipitation and lyophilization.
Development of highly stabilized curcumin nanoparticles by flash nanoprecipitation and lyophilization.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-06)
Shing Fung Chow, Ka Yee Wan, Kwok Kin Cheng, Ka Wai Wong, Changquan Calvin Sun, Larry Baum, Albert Hee Lum Chow
ABSTRACT
The influence of critical operating parameters on the Flash Nanoprecipitation (FNP) and resulting material properties of curcumin (CUR) nanoparticles has been evaluated using a confined impinging jets-with-dilution mixer (CIJ-D-M). It has been shown that the mixing rate, molecular weight of polymeric stabilizer (i.e., polyethylene glycol-b-poly(dl-lactide) di-block copolymer; PEG-PLA) and drug-to-copolymer mass ratio all exert a significant impact on the particle size and stability of the generated nanosuspensions. The attainable mean particle size and span of the nanoparticles through optimization of these process parameters were approximately 70nm and 0.85 respectively. However, the optimized nanosuspension was only stable for about two hours after preparation. Co-formulation with polyvinylpyrrolidone (PVP) substantially extended the product lifespan to 5days at ambient conditions and two weeks at 4°C. Results from zeta potential measurement and X-ray photoelectron spectroscopy (XPS) suggested that the enhanced stability is probably due to the formation of an additional protective barrier by PVP around the particle surface, thereby suppressing the dissociation of PEG-PLA from the particles and preventing CUR leakage from inside. Long-term storage stability (>1year) could be achieved by lyophilization of the optimized nanosuspension with Kleptose (hydroxypropyl-β-cyclodextrin), which was shown to be the only effective lyoprotectant among all the ones tested for the CUR nanoparticles. At an optimal concentration of Kleptose (1.25% w/v), the redispersibility (Sf/Si; ratio of the final and initial particle sizes) and encapsulation efficiency of lyophilized CUR nanoparticles were about 1.22% and 94%, respectively.
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from starch
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥99% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-(+)-Trehalose dihydrate, ≥98.5% (HPLC), from Saccharomyces cerevisiae
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)