Merck
  • Identification and characterization of enolase as a collagen-binding protein in Lactobacillus plantarum.
Identification and characterization of enolase as a collagen-binding protein in Lactobacillus plantarum.
Journal of basic microbiology (2015-02-28)
Marzia Salzillo, Valeria Vastano, Ugo Capri, Lidia Muscariello, Margherita Sacco, Rosangela Marasco
ABSTRACT
Collagen is a target of pathogens for adhesion, colonization, and invasion of host tissue. Probiotic bacteria can mimic the same mechanism as used by the pathogens in the colonization process, expressing cell surface proteins that specifically interact with extracellular matrix component proteins. The capability to bind collagen is expressed by several Lactobacillus isolates, including some Lactobacillus plantarum strains. In this study we report the involvement of the L. plantarum EnoA1 alfa-enolase in type I collagen (CnI) binding. By adhesion assays, we show that the mutant strain LM3-CC1, carrying a null mutation in the enoA1 gene, binds to immobilized collagen less efficiently than wild type strain. CnI overlay assay and Elisa tests, performed on the purified EnoA1, show that this protein can bind collagen both under denaturing and native conditions. By using truncated recombinant enolase proteins, we also show that the region spanning from 73rd to the 140th amino acid residues is involved in CnI binding.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine