Merck
  • Physisorption-Based Charge Transfer in Two-Dimensional SnS2 for Selective and Reversible NO2 Gas Sensing.
Physisorption-Based Charge Transfer in Two-Dimensional SnS2 for Selective and Reversible NO2 Gas Sensing.
ACS nano (2015-10-09)
Jian Zhen Ou, Wanyin Ge, Benjamin Carey, Torben Daeneke, Asaf Rotbart, Wei Shan, Yichao Wang, Zhengqian Fu, Adam F Chrimes, Wojtek Wlodarski, Salvy P Russo, Yong Xiang Li, Kourosh Kalantar-Zadeh
ABSTRACT
Nitrogen dioxide (NO2) is a gas species that plays an important role in certain industrial, farming, and healthcare sectors. However, there are still significant challenges for NO2 sensing at low detection limits, especially in the presence of other interfering gases. The NO2 selectivity of current gas-sensing technologies is significantly traded-off with their sensitivity and reversibility as well as fabrication and operating costs. In this work, we present an important progress for selective and reversible NO2 sensing by demonstrating an economical sensing platform based on the charge transfer between physisorbed NO2 gas molecules and two-dimensional (2D) tin disulfide (SnS2) flakes at low operating temperatures. The device shows high sensitivity and superior selectivity to NO2 at operating temperatures of less than 160 °C, which are well below those of chemisorptive and ion conductive NO2 sensors with much poorer selectivity. At the same time, excellent reversibility of the sensor is demonstrated, which has rarely been observed in other 2D material counterparts. Such impressive features originate from the planar morphology of 2D SnS2 as well as unique physical affinity and favorable electronic band positions of this material that facilitate the NO2 physisorption and charge transfer at parts per billion levels. The 2D SnS2-based sensor provides a real solution for low-cost and selective NO2 gas sensing.
Sigma-Aldrich
Tin(IV) chloride, 99.995% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Tin(IV) chloride, 98%
Sigma-Aldrich
Tin(IV) chloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%