Merck
  • Determination of 3-mercaptopropionic acid by HPLC: A sensitive method for environmental applications.
Determination of 3-mercaptopropionic acid by HPLC: A sensitive method for environmental applications.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-15)
P Salgado, T Visnevschi-Necrasov, R P Kiene, I Azevedo, A C S Rocha, C M R Almeida, C Magalhães
ABSTRACT
The organic sulfur compound 3-mercaptopropionic acid (3-MPA) is an important thiol intermediate in organic sulfur metabolism in natural environments. It is generated during degradation of sulfur-containing amino acids (e.g. methionine) and from demethylation of dimethylsulfoniopropionate (DMSP). This pathway is an alternative enzymatic process in the DMSP catabolism that routes sulfur away from the climatically-active dimethyl sulfide (DMS). 3-MPA detection and subsequent quantification in different matrices is difficult due to its extreme reactivity. We therefore developed a sensitive method for determination of 3-MPA based on pre-column derivatization with monobromobimane and analysis by high-performance liquid chromatography (HPLC) with fluorescence detection. This methodology was first tested with 3-MPA standards under low (0.005-0.2μmolL(-1)) and high (1-25μmolL(-1)) concentrations. For the optimization of the reaction, CHES and, alternatively, Tris-HCl buffers were evaluated in the derivatization step, with Tris-HCl showing more effective separation of thiol derivatives and a better 3-MPA peak shape. The detection limit was 4.3nmolL(-1) with a 10μL sample injection, and mean recoveries of 3-MPA ranged from 97 to 105% in estuarine waters with different salinities (0.17 and 35.9ppt). The linearity (r>0.99) and repeatability of detector response, with intra- and inter-day precision (% CV) of 2.68-7.01% and 4.86-12.5%, respectively, confirmed the reliability of the method. Previous 3-MPA analytical methods required immediate analysis due to unstable derivatives, but in this method we achieved high stability of the derivatized samples when stored at 4°C, with only a 3-5% loss after more than one year of storage. This method was successfully applied to measure 3-MPA concentrations and rates of 3-MPA production in a variety of intertidal estuarine sediment slurries. Dissolved 3-MPA concentrations in these sediment slurries varied between 2 and 237μmolL(-1) and, 3-MPA net fluxes ranged in wet sediments between -3.6±1.7 and 30±5μmolL(-1)g(-1)h(-1). Thus, the application of this optimized methodology showed an efficient performance for measuring 3-MPA in environmental samples, with a straightforward sample derivatization and a simple analysis of stable 3-MPA derivatives.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
CHES, BioUltra, ≥99.5% (T)
Sigma-Aldrich
CHES, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
CHES, ≥99.0% (titration)
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.