Merck
  • Protection of human myeloid dendritic cell subsets against influenza A virus infection is differentially regulated upon TLR stimulation.
Protection of human myeloid dendritic cell subsets against influenza A virus infection is differentially regulated upon TLR stimulation.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-25)
Faezzah Baharom, Saskia Thomas, Andrea Bieder, Maria Hellmér, Julia Volz, Kerrie J Sandgren, Gerald M McInerney, Gunilla B Karlsson Hedestam, Ira Mellman, Anna Smed-Sörensen
ABSTRACT
The proinflammatory microenvironment in the respiratory airway induces maturation of both resident and infiltrating dendritic cells (DCs) upon influenza A virus (IAV) infection. This results in upregulation of antiviral pathways as well as modulation of endocytic processes, which affect the susceptibility of DCs to IAV infection. Therefore, it is highly relevant to understand how IAV interacts with and infects mature DCs. To investigate how different subsets of human myeloid DCs (MDCs) involved in tissue inflammation are affected by inflammatory stimulation during IAV infection, we stimulated primary blood MDCs and inflammatory monocyte-derived DCs (MDDCs) with TLR ligands, resulting in maturation. Interestingly, MDDCs but not MDCs were protected against IAV infection after LPS (TLR4) stimulation. In contrast, stimulation with TLR7/8 ligand protected MDCs but not MDDCs from IAV infection. The reduced susceptibility to IAV infection correlated with induction of type I IFNs. We found that differential expression of TLR4, TRIF, and MyD88 in the two MDC subsets regulated the ability of the cells to enter an antiviral state upon maturation. This difference was functionally confirmed using small interfering RNA and inhibitors. Our data show that different human MDC subsets may play distinct roles during IAV infection, as their capacity to induce type I IFNs is dependent on TLR-specific maturation, resulting in differential susceptibility to IAV infection.
Sigma-Aldrich
Sodium chloride solution, 0.85%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, tablet
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
SAFC
Sodium chloride solution, 5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
SAFC
HEPES
SKU
Pack Size
Availability
Price
Quantity
SAFC
HEPES
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture