Merck
  • Low-glycemic index diet may improve insulin sensitivity in obese children.
Low-glycemic index diet may improve insulin sensitivity in obese children.
Pediatric research (2015-08-14)
Chonnikant Visuthranukul, Pathama Sirimongkol, Aree Prachansuwan, Chandhita Pruksananonda, Sirinuch Chomtho
ABSTRACT
A low-glycemic index (GI) diet may be beneficial for weight management due to its effect on insulin metabolism and satiety. Obese children aged 9-16 y were randomly assigned either a low-GI diet or a low-fat diet (control group) for 6 mo. Body composition changes were measured by dual-energy X-ray absorptiometry and bioelectrical impedance analysis. Insulin sensitivity was measured by fasting plasma glucose and insulin. Fifty-two participants completed the study (mean age: 12.0 ± 2.0 y, 35 boys); both groups showed significantly decreased BMI z-score but similar changes in fat and fat-free mass. The low-GI group demonstrated a significant decline in fasting plasma insulin (22.2 ± 14.3 to 13.7 ± 10.9 mU/l; P = 0.004) and homeostatic model of assessment-insulin resistance (4.8 ± 3.3 to 2.9 ± 2.3; P = 0.007), whereas the control group did not. However, general linear model showed no significant difference in insulin resistance between groups after adjusting for baseline levels, suggesting that the greater reduction in insulin resistance in the low-GI group may be explained by higher baseline values. Despite subtle effects on body composition, a low-GI diet may improve insulin sensitivity in obese children with high baseline insulin. A bigger study in obese children with insulin resistance could be worthwhile to confirm our findings.
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
L-Alanine-12C3, 99.9 atom % 12C