Merck
  • Ultra high performance liquid chromatography with mass spectrometry method for the simultaneous determination of phenolic constituents in honey from various floral sources using multiwalled carbon nanotubes as extraction sorbents.
Ultra high performance liquid chromatography with mass spectrometry method for the simultaneous determination of phenolic constituents in honey from various floral sources using multiwalled carbon nanotubes as extraction sorbents.
Journal of separation science (2015-05-20)
Saikh Mohammad Wabaidur, Yacine Badjah Hadj Ahmed, Zeid Abdullah Alothman, Munir Saeed Obbed, Nasser Mohamed AL-Harbi, Turki Mohammad AL-Turki
ABSTRACT
An ultra high performance liquid chromatography with mass spectrometry method has been developed for the simultaneous separation, identification and determination of 22 phenolic constituents in honey from various floral sources from Yemen. Solid-phase extraction was used for extraction of the target phenolic constituents from honey samples, while multiwalled carbon nanotubes were used as solid-phase adsorbent. The chromatographic separation of all phenolic constituents was performed on a BEH C18 column using a linear gradient elution with a binary mobile phase mixture of aqueous 0.1% formic acid and methanol. The quantitation was carried out in selected ion reaction monitoring acquisition mode. The total amount of phenolic acids, flavonoids and other phenols in each analyzed honey was found in the range of 338-3312, 122-5482 and 2.4-1342 μg/100 g of honey, respectively. 4-Hydroxybenzoic acid was found to be the major phenolic acid. The main detected flavonoid was chrysin, while cinnamic acid was found to be the major other phenol compound. The regeneration of solid phase adsorbent to be reused and recovery results confirm that the proposed method could be potentially used for the routine analysis of phenolic constituents in honey extract.
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Galangin, autophagy inducing flavonoid
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C