Merck
  • Vitamins D and A can be successfully measured by LC-MS/MS in cord blood diluted plasma.
Vitamins D and A can be successfully measured by LC-MS/MS in cord blood diluted plasma.
Clinical biochemistry (2015-04-30)
Ali A Albarhani, Fiona Collier, Ronda F Greaves, Anne-Louise Ponsonby, Katrina J Allen, Peter J Vuillermin, Peter Roche, Michael W Clarke
ABSTRACT
In widely used protocols for the collection and isolation of cord blood mononuclear cells, investigators are left with substantial volumes of diluted plasma which could be used for other measurements. The aim of this study was to ascertain the validity of umbilical cord blood (UCB) diluted plasma samples for vitamin D, A and E analysis compared to UCB serum samples. Twenty UCB matched samples of diluted plasma and serum were collected. The samples were analysed by two liquid chromatography-tandem mass spectrometry (LC-MS/MS) methods on two separate occasions. The results of 25(OH)D3 obtained by the two laboratories demonstrated close agreement with a mean difference of 0.14nmol/L [95% confidence interval (95% CI), -6.8 to 7.1]. Both methods demonstrate close agreement for 25(OH)D3 in UCB serum versus diluted UCB plasma; mean difference 2.2nmol/L [95% CI, -9.5 to 13.9] and 4.1nmol/L [95% CI, -14.5 to 6.1] for the results from Lab A and Lab B, respectively. Vitamin A was quantified by Lab A in UCB serum and diluted UCB plasma; mean difference 0.07μmol/L [95% CI, -0.41 to 0.28]. Results of 25(OH)D3 epimer and vitamin E in the diluted UCB plasma were below the limit of quantification, and could not be compared with UCB serum. Diluted UCB plasma can be used for the quantification of retinol and 25(OH)D3 by LC-MS/MS. By contrast, quantification of 25(OH)D3 epimer and vitamin E in diluted UCB plasma is not supported by this study due to limitations in analytical sensitivity.
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Supelco
(±)-α-Tocopherol, analytical standard