Merck
  • Hypomethylation of DNA-binding inhibitor 4 serves as a potential biomarker in distinguishing acquired tamoxifen-refractory breast cancer.
Hypomethylation of DNA-binding inhibitor 4 serves as a potential biomarker in distinguishing acquired tamoxifen-refractory breast cancer.
International journal of clinical and experimental pathology (2015-10-16)
Yonghui Zhang, Bin Zhang, Jing Fang, Xuchen Cao
ABSTRACT
To explore the methylation status of DNA-binding inhibitor 4 (ID4) in tamoxifen-refractory (TR) breast cancer. From January 2012 to December 2014, breast cancer patients managed by radical mastectomy previously and receiving tamoxifen treatment for at least 12 months were enrolled. According to the response to tamoxifen, patients were divided into TR group and tamoxifen-sensitive (TS) group. Genomic DNA was isolated from fasting venous blood, and methylight technique was applied to determine the methylation status of ID4. 43 patients with TS breast cancer and 31 patients with TR breast cancer were enrolled. No significant difference between groups was observed in term of patients' characteristics, such as age (P=0.693), progesterone receptor (P=0.970), menopausal status (P=0.784) and histological type (P=0.537), while the stage of cancer in TR group was significantly higher than TS group (P<0.001). Compared to TS group, PMR of ID4 was significantly higher in TR group (P=0.002). ROC curve analysis indicated that ID4 yielded an AUC of 0.716 with 77.4% sensitivity and 62.79% specificity in distinguishing TR breast cancer at the cut point of 3.8%. The PMR cut point of ID4 was set at 6.8% in survival analysis, log-rank test indicated the risk of disease progression was comparable between patients with ID4 hypermethylation or hypomethylation (P=0.287). ID4 hypomethylation is present in TR breast cancer, and it may serve as a potential biomarker in distinguishing TR breast cancer. However, the results need further validation in larger studies.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%