Merck
  • Influence of pH, concentration and light on stability of allicin in garlic (Allium sativum L.) aqueous extract as measured by UPLC.
Influence of pH, concentration and light on stability of allicin in garlic (Allium sativum L.) aqueous extract as measured by UPLC.
Journal of the science of food and agriculture (2014-09-11)
Haiping Wang, Xixiang Li, Xinyan Liu, Di Shen, Yang Qiu, Xiaohui Zhang, Jiangping Song
ABSTRACT
Garlic is one of the most important bulb vegetables and is mainly used as a spice or flavoring agent for foods. It is also cultivated for its medicinal properties, attributable to sulfur compounds, of which allicin is the most important. However, the stability of allicin in garlic extract is not well understood. In this study, using UPLC, the stability of allicin extracted in water from garlic was evaluated in phosphate buffer at different temperatures under light and dark conditions. At room temperature, allicin in aqueous extract was most stable at pH 5-6 but degraded quickly at lower or higher pH. It began to degrade within 0.5 h and was not detectable after 2 h when the pH was higher than 11 or lower than 1.5. It degraded quickly when the temperature was higher than 40 °C and especially higher than 70 °C. At room temperature, allicin in water could be stored for 5 days without obvious degradation. Higher concentrations of allicin in solution were somewhat more stable than low concentrations. Allicin extract was sensitive to pH and temperature of storage but not to light. Higher-concentration allicin solution was more stable.
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri