Merck
  • Immunophenotypic analysis of CD103+ B-lymphoproliferative disorders: hairy cell leukemia and its mimics.
Immunophenotypic analysis of CD103+ B-lymphoproliferative disorders: hairy cell leukemia and its mimics.
American journal of clinical pathology (2009-03-18)
Henry Y Dong, James Weisberger, Zach Liu, Sorina Tugulea
ABSTRACT
CD103 is characteristically expressed in hairy cell leukemia (HCL), a B-lymphoproliferative disorder highly responsive to treatment with purine analogs. Other CD103+ diseases are rare and do not respond well to the same therapy, including HCL variant (HCLv) and splenic marginal zone B-cell lymphoma (SMZL) variants. We analyzed 215 cases of CD103+ B-lymphoproliferative disorders to further delineate their immunophenotypic features. Flow cytometric analysis revealed that 78.6% of all cases expressed CD25 and CD103, characteristic of classical HCL. Cases analyzed immunohistochemically were also invariably positive for annexin-A1; a subset coexpressed CD10 (33/169 [19.5%]) or BCL1 (26/65 [36.9%]). In contrast, 21.4% of cases lacked CD25, a subset of which was analyzed and was invariably negative for annexin-A1, CD10, and BCL1. The CD25- cases had variable morphologic features ranging from HCLv and SMZL to prolymphocytic leukemia and diffuse large B-cell lymphoma. Clinically, patients with CD25- disease tended to be older (P= .001), typically had leukocytosis (P= .014), and did not respond well to cladribine or pentostatin. We suggest categorizing CD103+ B-lymphoproliferative disorders into 2 groups. While HCL coexpresses CD25 and annexin-A1, diseases lacking CD25 and annexin-A1 behave clinically differently and can be separated from HCL on diagnosis.
Sigma-Aldrich
CD103 (EP206) Rabbit Monoclonal Primary Antibody