Merck
Recycled IL-7 Can Be Delivered to Neighboring T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2015-04-15)
Douglas A Bazdar, Magdalena Kalinowska, Soumya Panigrahi, Scott F Sieg
ABSTRACT
IL-7 is a key homeostatic cytokine that provides signals for T cell survival and proliferation in vivo. In this article, we provide evidence that IL-7 utilization is enhanced by a novel mechanism of cytokine "recycling" during which T cells treated with rIL-7 are rapidly induced to express p-STAT5 and are subsequently able to recycle biologically active cytokine for release to neighboring cells in soluble form. Our observations indicate that the ability of cells to recycle IL-7 is dependent on IL-7R α-chain (CD127) and endocytosis, consistent with a model whereby IL-7 is internalized via receptor interactions before recycling. These observations provide evidence of a novel mechanism that enables cells to optimally use IL-7.
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-IL7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O