Merck
  • Stereochemistry of 1-(4'-hydroxyphenyl)ethanol produced by hydroxylation of 4-ethylphenol by p-cresol methylhydroxylase.
Stereochemistry of 1-(4'-hydroxyphenyl)ethanol produced by hydroxylation of 4-ethylphenol by p-cresol methylhydroxylase.
The Biochemical journal (1984-12-01)
W McIntire, D J Hopper, J C Craig, E T Everhart, R V Webster, M J Causer, T P Singer
ABSTRACT
Enzymic hydroxylation of 4-ethylphenol by (a) Pseudomonas putida and (b) highly purified p-cresol methylhydroxylase gave optically active 1-(4'-hydroxyphenyl)-ethanol. The products were transformed into the phenolic methyl ethers and shown to contain 69.5% and 65.6%, respectively, of the (S)-(-)-isomer. The stereochemistry of the reaction is discussed in terms of three distinct steps occurring at the active site of the enzyme.
Sigma-Aldrich
4-Ethylphenol, ≥98%, FG