Merck
  • Novel SCRG1/BST1 axis regulates self-renewal, migration, and osteogenic differentiation potential in mesenchymal stem cells.
Novel SCRG1/BST1 axis regulates self-renewal, migration, and osteogenic differentiation potential in mesenchymal stem cells.
Scientific reports (2014-01-15)
Emiko Aomatsu, Noriko Takahashi, Shunsuke Sawada, Naoto Okubo, Tomokazu Hasegawa, Masayuki Taira, Hiroyuki Miura, Akira Ishisaki, Naoyuki Chosa
ABSTRACT
Human mesenchymal stem cells (hMSCs) remodel or regenerate various tissues through several mechanisms. Here, we identified the hMSC-secreted protein SCRG1 and its receptor BST1 as a positive regulator of self-renewal, migration, and osteogenic differentiation. SCRG1 and BST1 gene expression decreased during osteogenic differentiation of hMSCs. Intriguingly, SCRG1 maintained stem cell marker expression (Oct-4 and CD271/LNGFR) and the potentials of self-renewal, migration, and osteogenic differentiation, even at high passage numbers. Thus, the novel SCRG1/BST1 axis determines the fate of hMSCs by regulating their kinetic and differentiation potentials. Our findings provide a new perspective on methods for ex vivo expansion of hMSCs that maintain native stem cell potentials for bone-forming cell therapy.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
ProteoExtract® Subcellular Proteome Extraction Kit
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Focal Adhesion Kinase Inhibitor I, The Focal Adhesion Kinase Inhibitor I, also referenced under CAS 4506-66-5, controls the biological activity of Foca Adhesion Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-SCRG1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-BST1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.