Merck
  • Chemical reporters for monitoring RNA synthesis and poly(A) tail dynamics.
Chemical reporters for monitoring RNA synthesis and poly(A) tail dynamics.
Chembiochem : a European journal of chemical biology (2012-04-20)
Markus Grammel, Howard Hang, Nicholas K Conrad
ABSTRACT
A versatile "clickable" nucleoside: Metabolic labeling of cells is useful in studying the dynamics of biological molecules. N(6) pA can be utilized by all three mammalian RNA polymerases, as well as poly(A) polymerase. Because of its alkyne modification, RNA labeled with N(6) pA can be visualized and purified by using click chemistry.
Sigma-Aldrich
N6-Propargyl-ATP sodium salt