Merck
  • Electrospray ionization mass spectrometry of non-covalent complexes formed between N-alkylimidazolium-containing zwitterionic sulfonates and protonated bases.
Electrospray ionization mass spectrometry of non-covalent complexes formed between N-alkylimidazolium-containing zwitterionic sulfonates and protonated bases.
European journal of mass spectrometry (Chichester, England) (2014-01-01)
Anton Podjava, Svjatoslav Kistkin, Elina Ausekle, Elina Priede, Peteris Mekss, Andris Zicmanis
ABSTRACT
This paper describes non-covalent complexes between zwitterionic 3-(1-alkyl-3N-imidazolio)- propane-1-sulfonates and different amines. Electrospray ionization (ESI) mass spectrometry and collision- induced dissociation were used to measure the stability of such complexes in solution and in the gas phase. Generally, zwitterionic sulfonates formed more abundant complexes with protonated 5-methylcytosine (5-MCH) than with aliphatic amines. The results show that the association constants and half-dissociation threshold energies of these complexes nonlinearly depend on the alkyl chain length of the zwitterion. It is shown that the complexes with the lowest stability exist in acetonitrile solution or in the gas phase. The factors responsible for this complicated behavior are discussed. The structure of the complexes was investigated by quantum chemical calculations using molecular mechanics and density functional theory. Hydrogen bonding is proposed as the main type of interaction responsible for the stability of ion-zwitterion complexes. In summary, the information obtained in this study could be used for the development of the new derivatization reagents for some compounds containing amidinium groups, like 5-MCH, to increase selectivity of ESI-based methods.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Pyrrolidine, 99%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexylamine, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
5-Methylcytosine hydrochloride, ≥99%